Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002713.3 | NP_002704.1 | 3 | 725 | ||
NM_014110.4 | NP_054829.2 | 3 | 323 | |||
NM_138558.2 | NP_612568.1 | 3 | 605 | |||
XM_017001594.1 | XP_016857083.1 | 3 | 245 | |||
GenBank mRNA | AF061958.1 | - | 3 | 251 | ||
AF061959.1 | - | 3 | 406 | |||
AF126488.1 | - | 3 | 239 | |||
AK292077.1 | - | 3 | 251 | |||
AK297771.1 | - | 3 | 251 | |||
AK304619.1 | - | 3 | 251 | |||
AK307570.1 | - | 3 | 251 | |||
AK313814.1 | - | 3 | 231 | |||
AX775985.1 | - | 3 | 239 | |||
BC001597.1 | - | 2 | 131 | |||
BC013360.2 | - | 2 | 181 | 3633676 | ||
BC093017.1 | - | 3 | 243 | 30531222 | ||
BI459680.1 | - | 3 | 286 | |||
DA167043.1 | - | 3 | 323 | |||
U14575.1 | - | 3 | 712 |