Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190447.1 | NP_001177376.1 | 2 | 409 | ||
NM_002718.4 | NP_002709.2 | 3 | 2844 | |||
NM_181897.2 | NP_871626.1 | 2 | 699 | |||
XM_006713686.3 | XP_006713749.1 | 3 | 3255 | |||
XM_011512956.2 | XP_011511258.1 | 3 | 3019 | |||
GenBank mRNA | AK293012.1 | - | 3 | 2824 | ||
AK298059.1 | - | 2 | 338 | |||
AK299544.1 | - | 2 | 369 | |||
AK302163.1 | - | 1 | 107 | |||
AK314243.1 | - | 3 | 2772 | |||
AK316258.1 | - | 2 | 652 | |||
BC065531.1 | - | 3 | 2791 | 6161260 | ||
L07590.1 | - | 3 | 2731 | |||
L12146.1 | - | 2 | 624 |