Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006823.3 | NP_006814.1 | 3 | 537 | ||
NM_181839.2 | NP_862822.1 | 2 | 408 | |||
XM_017013620.1 | XP_016869109.1 | 4 | 412 | |||
XM_017013621.1 | XP_016869110.1 | 3 | 378 | |||
XM_017013622.1 | XP_016869111.1 | 2 | 143 | |||
GenBank mRNA | AK311857.1 | - | 2 | 93 | ||
BC022265.2 | - | 1 | 39 | 4288676 | ||
BI597731.1 | - | 3 | 346 | |||
CR457052.1 | - | 1 | 13 | |||
DB480496.1 | - | 2 | 190 | |||
S76965.1 | - | 3 | 546 |