Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197097.2 | NP_001184026.2 | 4 | 632 | ||
NM_001197098.1 | NP_001184027.1 | 3 | 274 | |||
NM_002771.3 | NP_002762.2 | 3 | 295 | |||
NM_007343.3 | NP_031369.2 | 3 | 504 | |||
XM_011517965.1 | XP_011516267.1 | 4 | 555 | |||
GenBank mRNA | AB298285.1 | - | 3 | 504 | ||
AB298286.1 | - | 3 | 274 | |||
AY052783.1 | - | 4 | 409 | |||
BC030238.1 | - | 3 | 294 | |||
BC069476.1 | - | 3 | 295 | 7262223 | ||
BC069494.1 | - | 3 | 295 | 7262235 | ||
D45417.1 | - | 3 | 282 | |||
X15505.1 | - | 3 | 295 | |||
X71345.1 | - | 3 | 322 | |||
X72781.1 | - | 3 | 325 |