Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031246.3 | NP_112536.2 | 3 | 737 | ||
GenBank mRNA | M20882.1 | - | 3 | 648 |