Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 5 | 1410 | ||
NM_001316339.1 | NP_001303268.1 | 5 | 1323 | |||
NM_002780.4 | NP_002771.2 | 6 | 1689 | |||
NM_213633.2 | NP_998798.1 | 5 | 1410 | |||
XM_017026996.1 | XP_016882485.1 | 7 | 1751 | |||
XM_017027000.1 | XP_016882489.1 | 6 | 1476 | |||
GenBank mRNA | AF289606.1 | - | 6 | 1030 | ||
AK075149.1 | - | 6 | 1304 | |||
BC008405.1 | - | 8 | 1548 | 4043484 | ||
BC063127.1 | - | 6 | 1557 | 4768860 | ||
M33665.1 | - | 4 | 830 | |||
M94891.1 | - | 6 | 1537 | |||
U18468.1 | - | 6 | 1456 | |||
U18469.1 | - | 5 | 1199 | |||
X17097.1 | - | 6 | 1543 |