Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113410.1 | NP_001106881.1 | 2 | 227 | ||
NM_002785.2 | NP_002776.3 | 3 | 593 | |||
NM_203287.1 | NP_976032.2 | 2 | 227 | |||
GenBank mRNA | BC020711.1 | - | 2 | 217 | 4732286 | |
M69245.1 | - | 3 | 555 | |||
U25988.1 | - | 2 | 223 |