Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002792.3 | NP_002783.1 | 2 | 344 | ||
GenBank mRNA | AF022815.1 | - | 2 | 213 | ||
AF054185.1 | - | 2 | 274 | |||
AK127210.1 | - | 1 | 672 | |||
AK312025.1 | - | 2 | 239 | |||
BC001895.1 | - | 2 | 277 | |||
BC004427.1 | - | 2 | 262 | 2819923 | ||
BC014999.1 | - | 2 | 272 | |||
BG575009.1 | - | 2 | 332 | |||
BI258951.1 | - | 2 | 344 | |||
BI906714.1 | - | 2 | 230 | |||
BT007165.1 | - | 2 | 189 | |||
GQ472224.1 | - | 2 | 262 | |||
HY039591.1 | - | 2 | 344 |