Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002819.4 | NP_002810.1 | 13 | 1375 | ||
NM_031990.3 | NP_114367.1 | 13 | 1354 | |||
NM_031991.3 | NP_114368.1 | 12 | 1297 | |||
XM_005259597.2 | XP_005259654.1 | 13 | 1878 | |||
XM_005259598.2 | XP_005259655.1 | 12 | 1800 | |||
GenBank mRNA | AB208910.1 | - | 10 | 3775 | ||
BC002397.2 | - | 13 | 1342 | 2821531 | ||
BC004383.1 | - | 12 | 1207 | 3615251 | ||
BC013694.1 | - | 13 | 1288 | 3863892 | ||
BC023219.1 | - | 13 | 1341 | |||
BT006819.1 | - | 12 | 1269 | |||
X60648.1 | - | 12 | 1219 | |||
X62006.1 | - | 12 | 1236 | |||
X65371.1 | - | 13 | 1293 | |||
X65372.1 | - | 13 | 1314 | |||
X66975.1 | - | 13 | 1356 |