Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321386.1 | NP_001308315.1 | 3, 4 | 423 | ||
NM_001321387.1 | NP_001308316.1 | 3, 4 | 725 | |||
NM_002825.5 | NP_002816.1 | 3, 4 | 660 | |||
XM_017012473.1 | XP_016867962.1 | 3, 4 | 543 | |||
GenBank mRNA | AK290488.1 | - | 3, 4 | 489 | ||
AK313424.1 | - | 3, 4 | 563 | |||
BC005916.1 | - | 3, 4 | 648 | 4248708 | ||
BI462078.1 | - | 3, 4 | 432 | |||
BT019692.1 | - | 2, 3 | 297 | |||
BX420480.2 | - | 3, 4 | 724 | |||
BX441769.2 | - | 3, 4 | 428 | |||
CN413530.1 | - | 3, 4 | 409 | |||
CR450338.1 | - | 2, 3 | 297 | |||
D90226.1 | - | 3, 4 | 421 | |||
M57399.1 | - | 3, 4 | 692 | |||
X52946.1 | - | 3, 4 | 548 |