Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018663.2 | NP_061133.1 | 2 | 276 | ||
GenBank mRNA | AF250136.1 | - | 2 | 276 | ||
AY044439.1 | - | 2 | 211 | |||
BC009836.1 | - | 4 | 695 | |||
BC073997.1 | - | 2 | 247 | 6171609 |