Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252039.1 | NP_001238968.1 | 3 | 582 | ||
NM_004583.3 | NP_004574.2 | 2 | 366 | |||
NM_201434.2 | NP_958842.1 | 3 | 483 | |||
XM_011525090.1 | XP_011523392.1 | 2 | 305 | |||
XM_011525091.1 | XP_011523393.1 | 2 | 429 | |||
GenBank mRNA | AF141304.1 | - | 1 | 195 | ||
AF498938.1 | - | 1 | 126 | |||
AK310234.1 | - | 3 | 526 | |||
AL553867.3 | - | 2 | 321 | |||
BC106039.1 | - | 2 | 325 | 4938169 | ||
BC114439.1 | - | 2 | 321 | 40083572 | ||
BM424166.1 | - | 3 | 423 | |||
BM550607.1 | - | 2 | 326 | |||
BP222378.1 | - | 2 | 368 | |||
BT019484.1 | - | 1 | 126 | |||
CR541901.1 | - | 1 | 126 | |||
U11293.1 | - | 1 | 201 | |||
U18420.1 | - | 2 | 261 |