Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005055.4 | NP_005046.2 | 7, 8 | 1381 | ||
NM_032645.4 | NP_116034.2 | 5, 6 | 1204 | |||
XM_005253042.3 | XP_005253099.1 | 6, 7 | 1285 | |||
XM_005253043.3 | XP_005253100.1 | 6, 7 | 1216 | |||
XM_011520252.1 | XP_011518554.1 | 7, 8 | 1424 | |||
GenBank mRNA | AF449218.1 | - | 7, 8 | 1381 | ||
BC004196.2 | - | 5, 6 | 1169 | 3530551 | ||
KU178300.1 | - | 5, 6 | 990 | |||
KU178301.1 | - | 7, 8 | 1303 | |||
KU178302.1 | - | 7, 8 | 1167 | |||
KU178303.1 | - | 6, 7 | 1113 | |||
KU178304.1 | - | 6, 7 | 1044 | |||
Z33905.1 | - | 7, 8 | 1374 |