Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002890.2 | NP_002881.1 | 18 | 2589 | ||
NM_022650.2 | NP_072179.1 | 18 | 1983 | |||
XM_011543525.1 | XP_011541827.1 | 18 | 3041 | |||
GenBank mRNA | AB209106.1 | - | 18 | 2589 | ||
AK300263.1 | - | 19 | 2090 | |||
AK300269.1 | - | 18 | 1916 | |||
AK300406.1 | - | 19 | 2009 | |||
AK312739.1 | - | 18 | 1968 | |||
BC033015.2 | - | 18 | 2577 | 4829173 | ||
BC107870.1 | - | 18 | 1969 | |||
CR749722.1 | - | 19 | 3099 | |||
M23379.1 | - | 18 | 2508 | |||
M23612.1 | - | 18 | 1958 |