Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005339.1 | NP_001005339.1 | 3, 4 | 323 | ||
NM_002925.3 | NP_002916.1 | 3, 4 | 272 | |||
GenBank mRNA | AF045229.2 | - | 3, 4 | 252 | ||
AF368902.1 | - | 2, 3 | 227 | |||
AF493934.1 | - | 2, 3 | 209 | |||
AK290773.1 | - | 3, 4 | 278 | |||
AL547312.3 | - | 3, 4 | 272 | |||
AV707839.1 | - | 3, 4 | 272 | |||
BC009361.2 | - | 3, 4 | 268 | 4301597 | ||
BF685740.1 | - | 3, 4 | 268 | |||
BG763741.1 | - | 3, 4 | 279 | |||
BM914660.1 | - | 322 | ||||
CR457008.1 | - | 2, 3 | 209 |