Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002939.3 | NP_002930.2 | 8 | 1271 | ||
NM_203383.1 | NP_976317.1 | 8 | 1203 | |||
NM_203384.1 | NP_976318.1 | 7 | 1030 | |||
NM_203385.1 | NP_976319.1 | 7 | 1098 | |||
NM_203386.2 | NP_976320.1 | 8 | 1161 | |||
NM_203387.2 | NP_976321.1 | 8 | 1229 | |||
NM_203388.2 | NP_976322.1 | 7 | 988 | |||
NM_203389.2 | NP_976323.1 | 7 | 1056 | |||
XM_011520255.1 | XP_011518557.1 | 8 | 1206 | |||
XM_011520257.2 | XP_011518559.1 | 8 | 1454 | |||
XM_011520258.2 | XP_011518560.1 | 8 | 1422 | |||
XM_011520259.2 | XP_011518561.1 | 7 | 971 | |||
XM_011520260.2 | XP_011518562.1 | 8 | 1130 | |||
XM_011520261.2 | XP_011518563.1 | 8 | 1204 | |||
XM_011520262.2 | XP_011518564.1 | 8 | 1387 | |||
XM_011520263.1 | XP_011518565.1 | 7 | 938 | |||
XM_017018106.1 | XP_016873595.1 | 8 | 1271 | |||
GenBank mRNA | AB209196.1 | - | 6 | 2521 | ||
AL161967.1 | - | 6 | 2206 | |||
BC000677.2 | - | 7 | 958 | 3349381 | ||
BC003506.1 | - | 7 | 971 | 2959514 | ||
BC011186.1 | - | 6 | 756 | |||
BC011500.1 | - | 7 | 909 | 4155804 | ||
BC014629.2 | - | 7 | 958 | 3029730 | ||
BC024037.1 | - | 7 | 971 | 2959514 | ||
BC047730.1 | - | 8 | 1250 | 5788220 | ||
M22414.1 | - | 7 | 919 | |||
M36717.1 | - | 8 | 1158 | |||
X13973.1 | - | 8 | 1158 |