Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000968.3 | NP_000959.2 | 10 | 1311 | ||
GenBank mRNA | AB208977.1 | - | 10 | 1288 | ||
AK125772.1 | - | 11 | 1742 | |||
AK130784.1 | - | 10 | 1302 | |||
AK223057.1 | - | 10 | 1302 | |||
AK291117.1 | - | 10 | 1320 | |||
AK291859.1 | - | 10 | 1302 | |||
AK297484.1 | - | 3 | 891 | |||
BC001365.2 | - | 10 | 1292 | 3051487 | ||
BC005817.2 | - | 1 | 1279 | 3688641 | ||
BC007748.2 | - | 1 | 1333 | 4098568 | ||
BC007996.1 | - | 1 | 1285 | 3546003 | ||
BC009888.2 | - | 10 | 1279 | 4025800 | ||
BC010151.2 | - | 10 | 1284 | 3353592 | ||
BC014653.1 | - | 10 | 1280 | 4636179 | ||
BC018840.2 | - | 10 | 1292 | |||
BC030000.1 | - | 1 | 164 | |||
BC040610.1 | - | 1 | 1287 | |||
BC066925.1 | - | 10 | 1302 | 5259747 | ||
BC095427.1 | - | 10 | 1295 | 30711782 | ||
CA417911.1 | - | 1 | 144 | |||
D23660.1 | - | 10 | 1302 | |||
L20868.2 | - | 10 | 1258 |