Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001020.5 | NP_001011.1 | 2 | 252 | ||
XM_005259137.2 | XP_005259194.1 | 2 | 252 | |||
GenBank mRNA | AK055869.1 | - | 2 | 180 | ||
AK315498.1 | - | 2 | 180 | |||
BC004324.2 | - | 2 | 178 | 3628799 | ||
BC007977.2 | - | 2 | 163 | 4310307 | ||
BC024970.1 | - | 1 | 148 | |||
BC071674.1 | - | 2 | 181 | 5394209 | ||
BQ421722.1 | - | 2 | 210 | |||
CB107158.1 | - | 2 | 230 | |||
CB242275.1 | - | 4 | 390 | |||
M60854.1 | - | 2 | 164 |