Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001020.5 | NP_001011.1 | 3 | 339 | ||
NM_001321111.1 | NP_001308040.1 | 3 | 288 | |||
XM_005259137.2 | XP_005259194.1 | 3 | 339 | |||
GenBank mRNA | AA890305.1 | - | 3 | 368 | ||
AK055869.1 | - | 2 | 2112 | |||
AK315498.1 | - | 3 | 267 | |||
BC004324.2 | - | 3 | 265 | 3628799 | ||
BC007977.2 | - | 3 | 250 | 4310307 | ||
BC024970.1 | - | 1 | 235 | |||
BC071674.1 | - | 3 | 268 | 5394209 | ||
BC095429.1 | - | 3 | 209 | |||
BQ421722.1 | - | 3 | 297 | |||
CB107158.1 | - | 3 | 317 | |||
CB242275.1 | - | 3 | 303 | |||
M60854.1 | - | 3 | 251 |