Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005619.4 | NP_005610.1 | 10 | 1767 | ||
NM_206900.2 | NP_996783.1 | 9 | 1548 | |||
NM_206901.2 | NP_996784.1 | 6 | 590 | |||
GenBank mRNA | AF004222.1 | - | 10 | 1666 | ||
AF004223.1 | - | 7 | 816 | |||
AF004224.1 | - | 6 | 558 | |||
AF038540.1 | - | 7 | 997 | |||
AI971392.1 | - | 2 | 556 | |||
AK291967.1 | - | 9 | 1529 | |||
AK314158.1 | - | 9 | 1531 | |||
BC073874.1 | - | 10 | 1709 | 5242827 |