Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002986.2 | NP_002977.1 | 2 | 243 | ||
GenBank mRNA | BC017850.1 | - | 2 | 169 | 4618679 | |
BF197516.1 | - | 2 | 210 | |||
CR457421.1 | - | 2 | 102 | |||
U46573.1 | - | 2 | 155 | |||
Z69291.1 | - | 2 | 157 | |||
Z75668.1 | - | 3 | 157 | |||
Z75669.1 | - | 2 | 134 |