Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201359.1 | NP_001188288.1 | 5 | 482 | ||
NM_005624.3 | NP_005615.2 | 5 | 485 | |||
XM_011528177.2 | XP_011526479.1 | 6 | 686 | |||
XM_017027119.1 | XP_016882608.1 | 6 | 652 | |||
XM_017027120.1 | XP_016882609.1 | 6 | 581 | |||
XM_017027121.1 | XP_016882610.1 | 6 | 527 | |||
XM_017027122.1 | XP_016882611.1 | 6 | 661 | |||
GenBank mRNA | BC125136.1 | - | 4 | 375 | 40119547 |