Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002993.3 | NP_002984.1 | 2 | 356 | ||
GenBank mRNA | AK311981.1 | - | 2 | 233 | ||
BC013744.1 | - | 2 | 191 | 4471095 | ||
BP204144.1 | - | 2 | 356 | |||
U81234.1 | - | 2 | 224 | |||
Y08770.1 | - | 1 | 50 |