Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302123.1 | NP_001289052.1 | 2 | 346 | ||
NM_005409.4 | NP_005400.1 | 2 | 346 | |||
GenBank mRNA | AF002985.1 | - | 2 | 223 | ||
AF030514.1 | - | 2 | 205 | |||
AF352781.1 | - | 2 | 214 | |||
AL703031.1 | - | 2 | 186 | |||
BC005292.1 | - | 2 | 196 | 3934139 | ||
BC012532.1 | - | 2 | 350 | 4096572 | ||
BC110986.1 | - | 2 | 205 | 5178174 | ||
BP289021.1 | - | 2 | 248 | |||
BT006787.1 | - | 2 | 136 | |||
U59286.1 | - | 2 | 158 | |||
U66096.1 | - | 2 | 181 | |||
Y15220.1 | - | 2 | 142 |