Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000609.6 | NP_000600.1 | 2 | 192 | ||
NM_001033886.2 | NP_001029058.1 | 2 | 192 | |||
NM_001178134.1 | NP_001171605.1 | 2 | 192 | |||
NM_001277990.1 | NP_001264919.1 | 2, 3 | 192 | |||
NM_199168.3 | NP_954637.1 | 2 | 192 | |||
GenBank mRNA | AK292628.1 | - | 2 | 195 | ||
AK311814.1 | - | 2 | 190 | |||
AU120056.1 | - | 2, 3 | 187 | |||
AY644456.1 | - | 2 | 149 | |||
AY874118.1 | - | 2 | 179 | |||
BC039893.1 | - | 2 | 170 | 5729604 | ||
CD172464.1 | - | 2 | 190 | |||
CR450283.1 | - | 2 | 100 | |||
DA266752.1 | - | 2 | 193 | |||
DQ345517.1 | - | 2 | 144 | |||
DQ345518.1 | - | 2 | 139 | |||
DQ345519.1 | - | 2 | 144 | |||
DQ345520.1 | - | 2 | 144 | |||
L36033.1 | - | 2 | 179 | |||
L36034.1 | - | 2 | 179 | |||
U16752.1 | - | 2 | 180 |