Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003007.4 | NP_002998.1 | 2 | 1251 | ||
GenBank mRNA | BC007096.1 | - | 3 | 1002 | 4251523 | |
BC055416.1 | - | 3 | 1002 | 6668871 | ||
BT007177.1 | - | 3 | 981 | |||
J04440.1 | - | 2 | 1167 |