Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243879.1 | NP_001230808.1 | 3 | 547 | ||
NM_004593.2 | NP_004584.1 | 4 | 681 | |||
XM_005247703.1 | XP_005247760.1 | 5 | 849 | |||
XM_011513072.2 | XP_011511374.1 | 5 | 3651 | |||
XM_017007033.1 | XP_016862522.1 | 4 | 1827 | |||
GenBank mRNA | AB209208.1 | - | 5 | 3638 | ||
AK094495.1 | - | 1 | 603 | |||
AK098191.1 | - | 4 | 468 | |||
AK125610.1 | - | 1 | 1351 | |||
AK301118.1 | - | 3 | 400 | |||
AK308267.1 | - | 3 | 425 | |||
AK315632.1 | - | 4 | 665 | |||
BC000160.2 | - | 4 | 548 | 2984771 | ||
BC000451.2 | - | 4 | 547 | 2821443 | ||
BC005898.1 | - | 4 | 532 | 4046349 | ||
CR749214.1 | - | 3 | 2708 | |||
U61267.1 | - | 4 | 526 | |||
U68063.1 | - | 4 | 534 |