Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172357.1 | NP_001165828.1 | 3 | 461 | ||
NM_001172410.1 | NP_001165881.1 | 3 | 461 | |||
NM_001317778.1 | NP_001304707.1 | 3 | 461 | |||
NM_001317779.1 | NP_001304708.1 | 2 | 302 | |||
NM_001317780.1 | NP_001304709.1 | 3 | 461 | |||
NM_003018.3 | NP_003009.2 | 3 | 461 | |||
XM_011544613.2 | XP_011542915.1 | 2 | 241 | |||
GenBank mRNA | AK058094.1 | - | 6 | 1153 | ||
AK298119.1 | - | 3 | 332 | |||
AK315742.1 | - | 3 | 335 | |||
BC005913.1 | - | 3 | 334 | 4043169 | ||
BF433929.1 | - | 4 | 503 | |||
BG535994.1 | - | 3 | 338 | |||
BG569816.1 | - | 3 | 338 | |||
BI490051.1 | - | 2 | 166 | |||
BI524154.1 | - | 3 | 317 | |||
BM993053.1 | - | 4 | 535 | |||
BQ027183.1 | - | 4 | 527 | |||
DB537043.1 | - | 1 | 92 | |||
DQ884411.1 | - | 3 | 303 | |||
J03517.1 | - | 3 | 330 | |||
J03553.1 | - | 3 | 477 | |||
KU178329.1 | - | 3 | 303 | |||
KU178330.1 | - | 3 | 303 |