Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172410.1 | NP_001165881.1 | 4, 5 | 591 | ||
NM_001317779.1 | NP_001304708.1 | 3, 4 | 432 | |||
NM_003018.3 | NP_003009.2 | 4, 5 | 591 | |||
XM_011544613.2 | XP_011542915.1 | 3, 4 | 371 | |||
GenBank mRNA | AK315742.1 | - | 4, 5 | 465 | ||
BC005913.1 | - | 4, 5 | 464 | 4043169 | ||
BI490051.1 | - | 3, 4 | 296 | |||
BM993053.1 | - | 2, 3 | 405 | |||
BQ027183.1 | - | 2, 3 | 397 | |||
J03517.1 | - | 4, 5 | 460 | |||
J03553.1 | - | 4, 5 | 607 | |||
KU178329.1 | - | 4, 5 | 433 |