Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003032.2 | NP_003023.1 | 7 | 1427 | ||
NM_173216.2 | NP_775323.1 | 8 | 1750 | |||
NM_173217.2 | NP_775324.1 | 7 | 1093 | |||
XM_005247719.1 | XP_005247776.1 | 7 | 1338 | |||
XM_005247720.1 | XP_005247777.1 | 7 | 1396 | |||
XM_006713734.1 | XP_006713797.1 | 9 | 2005 | |||
XM_011513085.1 | XP_011511387.1 | 9 | 2013 | |||
XM_011513086.1 | XP_011511388.1 | 7 | 1396 | |||
XM_017007065.1 | XP_016862554.1 | 10 | 2097 | |||
XM_017007066.1 | XP_016862555.1 | 7 | 1425 | |||
XM_017007067.1 | XP_016862556.1 | 7 | 1545 | |||
XM_017007068.1 | XP_016862557.1 | 7 | 1430 | |||
GenBank mRNA | A17362.1 | - | 8 | 1554 | ||
A23699.1 | - | 5 | 1131 | |||
AK128726.1 | - | 3 | 2041 | |||
AK292879.1 | - | 7 | 1448 | |||
AK312023.1 | - | 4 | 456 | |||
BC031476.1 | - | 7 | 1421 | 4339124 | ||
BC040009.1 | - | 7 | 1427 | 5557868 | ||
X17247.1 | - | 8 | 1548 | |||
X54363.1 | - | 7 | 1386 | |||
X62822.1 | - | 7 | 1427 |