Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006931.2 | NP_008862.1 | 10 | 1581 | ||
GenBank mRNA | AB209607.1 | - | 9 | 2060 | ||
AK074861.1 | - | 2 | 222 | |||
AK298673.1 | - | 9 | 1278 | |||
AK304509.1 | - | 10 | 1284 | |||
AK312386.1 | - | 10 | 1472 | |||
AY034634.1 | - | 4 | 418 | |||
BC039196.1 | - | 10 | 1456 | 4396508 | ||
CR457413.1 | - | 9 | 1319 | |||
M20681.1 | - | 10 | 1561 |