Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007468.2 | NP_001007469.1 | 7 | 1050 | ||
NM_001317946.1 | NP_001304875.1 | 7 | 1104 | |||
NM_003073.4 | NP_003064.2 | 7 | 1077 | |||
XM_011530345.2 | XP_011528647.1 | 7 | 1100 | |||
GenBank mRNA | AB017523.1 | - | 7 | 811 | ||
AJ011737.1 | - | 7 | 985 | |||
AJ011738.1 | - | 7 | 958 | |||
AK021419.1 | - | 7 | 983 | |||
AK024025.1 | - | 7 | 1072 | |||
BC117114.1 | - | 7 | 866 | 40125665 | ||
BC143667.1 | - | 7 | 866 | 9052181 | ||
CB161660.1 | - | 5 | 550 | |||
CR456581.1 | - | 7 | 891 |