Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003801.1 | NP_001003801.1 | 10 | 1161 | ||
NM_001003802.1 | NP_001003802.1 | 11 | 1150 | |||
NM_003078.3 | NP_003069.2 | 11 | 1189 | |||
XM_011516521.2 | XP_011514823.1 | 10 | 1268 | |||
XM_017012556.1 | XP_016868045.1 | 10 | 1004 | |||
GenBank mRNA | AK127595.1 | - | 11 | 1697 | ||
AY450430.1 | - | 10 | 1465 | |||
AY450431.1 | - | 11 | 1150 | |||
BC002628.2 | - | 11 | 1150 | 3608512 | ||
BC110350.1 | - | 10 | 1080 | 5212363 | ||
BX648385.1 | - | 9 | 1912 |