Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003801.1 | NP_001003801.1 | 11, 12 | 1398 | ||
NM_001003802.1 | NP_001003802.1 | 12, 13 | 1387 | |||
NM_003078.3 | NP_003069.2 | 12, 13 | 1426 | |||
XM_011516521.2 | XP_011514823.1 | 11, 12 | 1505 | |||
XM_017012556.1 | XP_016868045.1 | 11, 12 | 1241 | |||
GenBank mRNA | AK127595.1 | - | 12, 13 | 1934 | ||
AY450430.1 | - | 11, 12 | 1702 | |||
AY450431.1 | - | 12, 13 | 1387 | |||
BC002628.2 | - | 12, 13 | 1387 | 3608512 | ||
BC110350.1 | - | 11, 12 | 1317 | 5212363 | ||
BX648385.1 | - | 10, 11 | 2149 | |||
U66619.1 | - | 12, 13 | 1435 |