Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003092.4 | NP_003083.1 | 3 | 423 | ||
NM_198220.2 | NP_937863.1 | 3 | 323 | |||
GenBank mRNA | AB451234.1 | - | 2 | 187 | ||
AB451359.1 | - | 2 | 187 | |||
AK313004.1 | - | 3 | 357 | |||
BC008311.1 | - | 3 | 328 | |||
BC018022.1 | - | 3 | 375 | 4286926 | ||
BC036737.2 | - | 3 | 282 | 5533477 | ||
BF243328.1 | - | 401 | ||||
DA088017.1 | - | 3 | 323 | |||
M15841.1 | - | 3 | 312 |