Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004597.6 | NP_004588.1 | 3 | 325 | ||
NM_177542.3 | NP_808210.2 | 3 | 289 | |||
GenBank mRNA | AK291912.1 | - | 2 | 104 | ||
BC000486.2 | - | 2 | 96 | 2819767 | ||
BC001930.2 | - | 1 | 74 | 2819767 | ||
BG528418.1 | - | 220 | ||||
BU531743.1 | - | 3 | 214 | |||
BU532756.1 | - | 370 | ||||
CB136390.1 | - | 2 | 515 | |||
CF130080.1 | - | 3 | 325 | |||
DA325073.1 | - | 3 | 217 | |||
U15008.1 | - | 2 | 100 |