Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080547.1 | NP_001074016.1 | 5 | 977 | ||
NM_003120.2 | NP_003111.2 | 5 | 974 | |||
XM_011520307.1 | XP_011518609.1 | 7 | 2419 | |||
XM_017018173.1 | XP_016873662.1 | 7 | 2422 | |||
GenBank mRNA | BI519776.1 | - | 1 | 378 | ||
BM561863.1 | - | 1 | 212 | |||
CD368431.1 | - | 2 | 418 | |||
X52056.1 | - | 5 | 944 |