Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204193.1 | NP_001191122.1 | 3 | 332 | ||
NM_001204196.1 | NP_001191125.1 | 2 | 256 | |||
NM_001204199.1 | NP_001191128.1 | 3 | 332 | |||
NM_003135.2 | NP_003126.1 | 3 | 332 | |||
GenBank mRNA | AK311803.1 | - | 3 | 240 | ||
BC000186.1 | - | 3 | 224 | |||
BC010947.1 | - | 3 | 216 | 4285753 | ||
BC017830.1 | - | 3 | 215 | 4252554 | ||
BU661702.1 | - | 2 | 134 | |||
BY798924.2 | - | 3 | 344 | |||
DA930867.1 | - | 3 | 215 | |||
X12791.1 | - | 3 | 224 |
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BE047584.1 | - | 6 | 357 |