Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000349.2 | NP_000340.2 | 4 | 588 | ||
XM_006716392.2 | XP_006716455.1 | 4 | 771 | |||
GenBank mRNA | AF035277.1 | - | 4 | 436 | ||
AK090513.1 | - | 3 | 772 | |||
AK315122.1 | - | 4 | 458 | |||
AL526126.3 | - | 4 | 595 | |||
AV708514.1 | - | 2 | 76 | |||
BC010550.1 | - | 4 | 389 | 4153601 | ||
CR456803.1 | - | 4 | 324 | |||
CR541850.1 | - | 4 | 324 | |||
KU178337.1 | - | 4 | 324 | |||
KU178338.1 | - | 4 | 324 | |||
S79669.1 | - | 4 | 476 | |||
U17280.1 | - | 4 | 450 |