Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009181.1 | NP_001009181.1 | 2 | 139 | ||
NM_003154.2 | NP_003145.1 | 2 | 139 | |||
GenBank mRNA | AK311812.1 | - | 2 | 104 | ||
BC067219.1 | - | 2 | 105 | 30328926 | ||
BX649104.1 | - | 1 | 577 | |||
CB958641.1 | - | 2 | 106 | |||
CB986973.1 | - | 143 | ||||
M18078.1 | - | 2 | 101 | |||
M18371.1 | - | 2 | 85 |