Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003190.4 | NP_003181.3 | 4 | 1058 | ||
NM_172208.2 | NP_757345.2 | 4 | 1058 | |||
NM_172209.2 | NP_757346.2 | 3 | 797 | |||
XM_011514828.2 | XP_011513130.1 | 4 | 925 | |||
XM_017011227.1 | XP_016866716.1 | 4 | 925 | |||
GenBank mRNA | AB010639.1 | - | 4 | 725 | ||
AF009510.1 | - | 4 | 728 | |||
AF029750.1 | - | 4 | 750 | |||
AF067286.3 | - | 4 | 720 | |||
AF314222.1 | - | 4 | 712 | |||
BC080574.1 | - | 4 | 883 | 6572933 | ||
EU693375.1 | - | 3 | 451 | |||
Y13582.1 | - | 4 | 841 |