Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TAZ SIRNA Assays ›
Gene Symbol:
TAZ
Gene Name:
tafazzin
Gene Aliases:
BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1
Chromosome Location:
Chr.X: 154411406 - 154421726 on Build GRCh38
UniGene ID:
Hs.409911
Species:
Human
siRNA ID
HSS144199
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TAZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000116.4
NP_000107.1
11
1122
NM_001303465.1
NP_001290394.1
10
1134
NM_181311.3
NP_851828.1
10
1032
NM_181312.3
NP_851829.1
10
1080
NM_181313.3
NP_851830.1
9
990
NR_024048.2
10
1138
XM_006724836.1
XP_006724899.1
11
1153
XM_006724837.1
XP_006724900.1
8
1138
XM_006724839.1
XP_006724902.1
9
1021
XM_006724841.3
XP_006724904.1
11
874
XM_006724842.3
XP_006724905.1
10
784
XM_011531189.1
XP_011529491.1
8
940
XM_011531191.2
XP_011529493.1
10
1312
XM_017029761.1
XP_016885250.1
8
1084
XM_017029762.1
XP_016885251.1
10
1063
XM_017029763.1
XP_016885252.1
8
886
XM_017029764.1
XP_016885253.1
9
651
XM_017029765.1
XP_016885254.1
9
1189
GenBank mRNA
AK130900.1
-
5
1485
AK291848.1
-
11
990
AY231461.1
-
10
714
AY231462.1
-
10
762
AY231463.1
-
9
672
AY231464.1
-
11
804
AY258036.1
-
11
804
AY258037.1
-
9
672
AY258038.1
-
10
762
AY258039.1
-
10
714
BC005062.1
-
10
1095
BC011515.1
-
7
803
4339983
DQ884408.1
-
10
808
KU178348.1
-
7
706
KU178349.1
-
11
796
KU178350.1
-
10
754
KU178351.1
-
9
751
KU178352.1
-
10
706
KU178353.1
-
9
664
X92762.1
-
11
1084
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
heart development
skeletal muscle tissue development
hemopoiesis
cardiolipin biosynthetic process
mitochondrial respiratory chain complex I assembly
cardiolipin acyl-chain remodeling
cristae formation
mitochondrial ATP synthesis coupled electron transport
cardiac muscle tissue development
cardiac muscle contraction
1-acylglycerol-3-phosphate O-acyltransferase activity
O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-nlx55:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline