Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003218.3 | NP_003209.2 | 9 | 1930 | ||
NM_017489.2 | NP_059523.2 | 10 | 1990 | |||
XM_005251291.3 | XP_005251348.1 | 11 | 2078 | |||
XM_005251292.3 | XP_005251349.1 | 10 | 2018 | |||
XM_011517582.2 | XP_011515884.1 | 12 | 2145 | |||
XM_017013792.1 | XP_016869281.1 | 10 | 1828 | |||
GenBank mRNA | AF003001.1 | - | 9 | 1915 | ||
AK123410.1 | - | 3 | 2201 | |||
CX788828.1 | - | 1 | 595 | |||
U40705.1 | - | 10 | 1975 | |||
U74382.1 | - | 9 | 1900 |