Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003274.4 | NP_003265.3 | 5, 6 | 851 | ||
XM_005261168.4 | XP_005261225.1 | 5, 6 | 844 | |||
XM_011529714.2 | XP_011528016.1 | 5, 6 | 844 | |||
XM_011529715.2 | XP_011528017.1 | 5, 6 | 844 | |||
XM_011529716.2 | XP_011528018.1 | 5, 6 | 844 | |||
XM_011529717.2 | XP_011528019.1 | 5, 6 | 844 | |||
XM_011529718.2 | XP_011528020.1 | 5, 6 | 844 | |||
XM_011529720.2 | XP_011528022.1 | 5, 6 | 844 | |||
XM_011529721.2 | XP_011528023.1 | 2, 3 | 217 | |||
XM_011529722.2 | XP_011528024.1 | 2, 3 | 183 | |||
XM_011529725.2 | XP_011528027.1 | 5, 6 | 844 | |||
XM_011529726.2 | XP_011528028.1 | 5, 6 | 844 | |||
XM_011529728.2 | XP_011528030.1 | 2, 3 | 361 | |||
XM_011529729.2 | XP_011528031.1 | 2, 3 | 238 | |||
XM_011529730.2 | XP_011528032.1 | 5, 6 | 844 | |||
XM_017028454.1 | XP_016883943.1 | 5, 6 | 844 | |||
GenBank mRNA | BC046241.1 | - | 5, 6 | 829 | ||
BC052247.1 | - | 5, 6 | 829 | |||
BC094823.1 | - | 5, 6 | 829 | |||
BC101728.1 | - | 5, 6 | 771 | 8069234 | ||
BX370317.2 | - | 2, 3 | 331 | |||
U19252.1 | - | 5, 6 | 819 | |||
U61500.1 | - | 5, 6 | 835 |