Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308394.1 | NP_001295323.1 | 3 | 593 | ||
NM_003278.2 | NP_003269.2 | 3 | 690 | |||
XM_017007116.1 | XP_016862605.1 | 2 | 651 | |||
XM_017007117.1 | XP_016862606.1 | 2 | 1216 | |||
GenBank mRNA | AK312093.1 | - | 3 | 698 | ||
BC011024.1 | - | 3 | 655 | 4042921 | ||
BI826174.1 | - | 3 | 592 | |||
BI826652.1 | - | 3 | 559 | |||
BM669340.1 | - | 1 | 176 | |||
CR541981.1 | - | 3 | 594 | |||
CR749292.1 | - | 3 | 4572 | |||
X64559.1 | - | 3 | 687 |