Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003279.2 | NP_003270.1 | 4 | 319 | ||
XM_011529031.2 | XP_011527333.1 | 4 | 329 | |||
GenBank mRNA | AI080746.1 | - | 3 | 373 | ||
AK291323.1 | - | 4 | 317 | |||
AK300665.1 | - | 317 | ||||
BC005323.1 | - | 4 | 317 | 3934188 | ||
BU854061.1 | - | 4 | 350 | |||
CR541864.1 | - | 3 | 253 | |||
CR541884.1 | - | 3 | 253 | |||
M22307.1 | - | 4 | 300 | |||
X07898.1 | - | 4 | 317 |