Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003279.2 | NP_003270.1 | 5, 6 | 506 | ||
XM_011529031.2 | XP_011527333.1 | 5, 6 | 516 | |||
GenBank mRNA | AI080746.1 | - | 1, 2 | 186 | ||
AK291323.1 | - | 5, 6 | 504 | |||
BC005323.1 | - | 5, 6 | 504 | 3934188 | ||
CR541864.1 | - | 4, 5 | 440 | |||
CR541884.1 | - | 4, 5 | 440 | |||
M22307.1 | - | 5, 6 | 487 | |||
X07898.1 | - | 5, 6 | 504 |