Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126132.2 | NP_001119604.1 | 11 | 775 | ||
NM_001126133.2 | NP_001119605.1 | 10 | 742 | |||
NM_001291774.1 | NP_001278703.1 | 11 | 837 | |||
NM_003283.5 | NP_003274.3 | 11 | 775 | |||
XM_011527246.2 | XP_011525548.1 | 10 | 652 | |||
XM_017027186.1 | XP_016882675.1 | 12 | 848 | |||
XM_017027187.1 | XP_016882676.1 | 11 | 836 | |||
GenBank mRNA | AK291352.1 | - | 11 | 627 | ||
AK308199.1 | - | 10 | 594 | |||
AY762903.1 | - | 10 | 570 | |||
AY762904.1 | - | 9 | 537 | |||
BC010963.1 | - | 11 | 628 | 4291828 | ||
BC022086.2 | - | 11 | 676 | |||
BC034143.1 | - | 11 | 627 | 4328089 | ||
BC107798.1 | - | 10 | 597 | 4247379 | ||
BT019630.1 | - | 9 | 537 | |||
M19308.1 | - | 10 | 595 | |||
M19309.1 | - | 11 | 627 | |||
S51938.1 | - | 9 | 536 | |||
S69208.1 | - | 11 | 611 | |||
S69209.1 | - | 10 | 578 |