Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000364.3 | NP_000355.2 | 10 | 487 | ||
NM_001001430.2 | NP_001001430.1 | 9 | 457 | |||
NM_001001431.2 | NP_001001431.1 | 9 | 457 | |||
NM_001001432.2 | NP_001001432.1 | 8 | 442 | |||
NM_001276345.1 | NP_001263274.1 | 10 | 487 | |||
NM_001276347.1 | NP_001263276.1 | 9 | 590 | |||
XM_006711508.3 | XP_006711571.1 | 9 | 487 | |||
XM_006711509.3 | XP_006711572.1 | 9 | 582 | |||
XM_011509938.2 | XP_011508240.1 | 10 | 519 | |||
XM_011509939.1 | XP_011508241.1 | 10 | 531 | |||
XM_011509940.2 | XP_011508242.1 | 10 | 535 | |||
XM_011509941.2 | XP_011508243.1 | 10 | 532 | |||
XM_011509942.2 | XP_011508244.1 | 8 | 489 | |||
XM_011509943.2 | XP_011508245.1 | 8 | 473 | |||
XM_011509944.2 | XP_011508246.1 | 8 | 486 | |||
XM_011509946.1 | XP_011508248.1 | 9 | 560 | |||
XM_017002216.1 | XP_016857705.1 | 9 | 465 | |||
XM_017002217.1 | XP_016857706.1 | 9 | 650 | |||
GenBank mRNA | AJ709280.1 | - | 9 | 477 | ||
AK125236.1 | - | 8 | 1604 | |||
AK290621.1 | - | 9 | 652 | |||
AK309493.1 | - | 3 | 291 | |||
AY277394.1 | - | 7 | 400 | |||
BC002653.2 | - | 9 | 423 | 3612968 | ||
L40162.1 | - | 9 | 418 | |||
S64668.1 | - | 8 | 377 | |||
X74819.1 | - | 8 | 415 | |||
X79856.1 | - | 9 | 395 | |||
X79859.1 | - | 7 | 619 | |||
X79861.1 | - | 8 | 407 |