Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243891.1 | NP_001230820.1 | 7 | 670 | ||
NM_001243892.1 | NP_001230821.1 | 6 | 616 | |||
NM_001243894.1 | NP_001230823.1 | 5 | 583 | |||
NM_001243895.1 | NP_001230824.1 | 6 | 696 | |||
NM_003288.3 | NP_003279.2 | 7 | 745 | |||
NM_199359.2 | NP_955391.1 | 6 | 685 | |||
NM_199360.2 | NP_955392.1 | 9 | 814 | |||
NM_199361.2 | NP_955393.1 | 8 | 754 | |||
NM_199362.2 | NP_955394.1 | 8 | 787 | |||
NM_199363.2 | NP_955395.1 | 7 | 727 | |||
NR_045090.1 | 7 | 714 | ||||
GenBank mRNA | AB062396.1 | - | 3 | 203 | ||
AF004429.1 | - | 4 | 192 | |||
AF004430.1 | - | 7 | 683 | |||
AK055068.1 | - | 7 | 686 | |||
AK125709.1 | - | 1 | 640 | |||
AK128744.1 | - | 2 | 2439 | |||
AK223031.1 | - | 7 | 657 | |||
AK293349.1 | - | 7 | 567 | |||
AK297064.1 | - | 6 | 643 | |||
AK298366.1 | - | 6 | 601 | |||
AK314501.1 | - | 7 | 703 | |||
BC006804.1 | - | 7 | 630 | 3446037 | ||
BI667351.1 | - | 8 | 566 | |||
BM473873.1 | - | 7 | 702 | |||
BQ948007.1 | - | 7 | 640 | |||
CR542036.1 | - | 7 | 607 | |||
CR749208.1 | - | 6 | 584 | |||
DA689198.1 | - | 5 | 482 |